Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133798.3 | NP_598559.2 | 5 | 1284 | ||
GenBank mRNA | AK146901.1 | - | 5 | 1165 | ||
AK147922.1 | - | 5 | 1145 | |||
AK158201.1 | - | 5 | 1175 | |||
AK162206.1 | - | 4 | 1052 | |||
AK164329.1 | - | 5 | 1160 | |||
AK167692.1 | - | 5 | 1285 | |||
BC018508.1 | - | 4 | 827 | 3156447 |