Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133800.3 | NP_598561.2 | 6 | 642 | ||
GenBank mRNA | AK036891.1 | - | 6 | 585 | ||
AK037035.1 | - | 6 | 636 | |||
AK083539.1 | - | 644 | ||||
AK167446.1 | - | 6 | 636 | |||
AY940114.1 | - | 6 | 585 | |||
BC013701.1 | - | 6 | 627 | 3483364 | ||
BC094424.1 | - | 6 | 629 | 6819926 |