Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133807.1 | NP_598568.1 | 7 | 1573 | ||
GenBank mRNA | AK053450.1 | - | 7 | 1600 | ||
AK150916.1 | - | 2 | 885 | |||
AK159374.1 | - | 7 | 1629 | |||
AK159552.1 | - | 7 | 1623 | |||
AK166576.1 | - | 7 | 1622 | |||
AK167604.1 | - | 7 | 1564 | |||
AK169250.1 | - | 5 | 1185 | |||
AK169390.1 | - | 7 | 1572 | |||
BC006877.1 | - | 7 | 1573 | 2647513 | ||
BC066172.1 | - | 7 | 1567 | 30429192 |