Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133809.1 | NP_598570.1 | 6 | 440 | ||
GenBank mRNA | AK129011.1 | - | 453 | |||
AK133074.1 | - | 548 | ||||
AK158251.1 | - | 6 | 636 | |||
BC014683.1 | - | 6 | 440 | 4210278 |