Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133810.3 | NP_598571.2 | 8 | 2016 | ||
GenBank mRNA | AK029551.1 | - | 8 | 1959 | ||
AK035693.1 | - | 8 | 1984 | |||
AK041139.1 | - | 4 | 1351 | |||
AK153108.1 | - | 8 | 1960 | |||
AK165291.1 | - | 8 | 1956 | |||
AY092028.1 | - | 8 | 1969 | |||
BC006579.1 | - | 8 | 1972 | 3586767 |