Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133832.3 | NP_598593.1 | 6 | 1951 | ||
GenBank mRNA | AK014244.1 | - | 2 | 1619 | ||
AK149656.1 | - | 6 | 2022 | |||
BC019796.1 | - | 6 | 1952 | 4923005 |