Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133847.3 | NP_598608.2 | 2, 3 | 378 | ||
GenBank mRNA | AB093220.2 | - | 1, 2 | 122 | ||
AK035622.1 | - | 2, 3 | 340 | |||
AK047497.1 | - | 2, 3 | 132 | |||
AK048158.1 | - | 2, 3 | 136 | |||
AK083541.1 | - | 2, 3 | 378 | |||
AK087048.1 | - | 2, 3 | 131 | |||
AK088012.1 | - | 2, 3 | 134 | |||
AK162435.1 | - | 2, 3 | 139 | |||
AK171487.1 | - | 2, 3 | 136 | |||
BC110309.1 | - | 2, 3 | 134 | 5324218 | ||
BC116320.2 | - | 2, 3 | 374 | 40096002 |