Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197082.1 | NP_001184011.1 | 11 | 1728 | ||
NM_133878.3 | NP_598639.1 | 10 | 1689 | |||
GenBank mRNA | AK004322.1 | - | 1 | 325 | ||
AK076089.1 | - | 10 | 1689 | |||
AK150153.1 | - | 10 | 1641 | |||
AK150610.1 | - | 10 | 1639 | |||
AK153193.1 | - | 10 | 1639 | |||
AK155225.1 | - | 7 | 2375 | |||
BC019807.1 | - | 10 | 1601 | 4456795 | ||
BC057645.1 | - | 11 | 1683 | 3591859 |