Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133887.4 | NP_598648.1 | 3 | 347 | ||
GenBank mRNA | AB019211.1 | - | 3 | 290 | ||
AK076390.1 | - | 332 | ||||
AK077678.1 | - | 3 | 348 | |||
AK078948.1 | - | 3 | 307 | |||
AK132618.1 | - | 3 | 311 | |||
AK146544.1 | - | 3 | 352 | |||
AK146735.1 | - | 3 | 339 | |||
AK167087.1 | - | 3 | 261 | |||
AK167804.1 | - | 3 | 311 | |||
AK168753.1 | - | 3 | 305 | |||
AK170203.1 | - | 3 | 384 | |||
AK187081.1 | - | 2 | 152 | |||
BC010669.1 | - | 3 | 334 | 2645010 |