Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133889.2 | NP_598650.2 | 11 | 1483 | ||
GenBank mRNA | AK004358.1 | - | 1 | 65 | ||
AK037201.1 | - | 11 | 1479 | |||
AK144053.1 | - | 11 | 1478 | |||
BC021480.1 | - | 10 | 1463 | |||
BC049111.1 | - | 11 | 1483 | 6415574 |