Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133907.3 | NP_598668.1 | 3 | 409 | ||
GenBank mRNA | AK029973.1 | - | 3 | 219 | ||
AK045211.1 | - | 3 | 206 | |||
AK046056.1 | - | 3 | 410 | |||
AK122187.1 | - | 3 | 344 | |||
AK140491.1 | - | 3 | 166 | |||
BC059245.1 | - | 3 | 225 | |||
BC120731.1 | - | 3 | 156 | 40129654 | ||
BC137626.1 | - | 3 | 156 | 8860645 |