Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133922.3 | NP_598683.3 | 14 | 2357 | ||
GenBank mRNA | AB041543.1 | - | 12 | 1989 | ||
AK029235.1 | - | 5 | 803 | |||
AK040525.1 | - | 14 | 2359 | |||
AK128931.1 | - | 14 | 2308 | |||
AK162934.1 | - | 14 | 2065 | |||
AK173281.1 | - | 14 | 4061 | |||
AK196294.1 | - | 1 | 146 | |||
BC066816.1 | - | 14 | 2277 | 30084911 |