Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pot1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133931.4 | NP_598692.1 | 9 | 1159 | ||
GenBank mRNA | AK036052.1 | - | 9 | 1160 | ||
AK040864.1 | - | 9 | 1152 | |||
AK085041.1 | - | 9 | 1150 | |||
AK135346.1 | - | 9 | 1129 | |||
AK137376.1 | - | 9 | 1129 | |||
BC016121.1 | - | 9 | 1121 | 4501733 |