Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133937.1 | NP_598698.1 | 2 | 181 | ||
GenBank mRNA | AK003342.1 | - | 2 | 196 | ||
AK020128.1 | - | 2 | 196 | |||
AK020277.1 | - | 2 | 197 | |||
AK150689.1 | - | 2 | 196 | |||
AK163236.1 | - | 2 | 196 | |||
AK166585.1 | - | 2 | 184 | |||
AK190298.1 | - | 2 | 120 | |||
AK193342.1 | - | 2 | 120 | |||
BC010582.1 | - | 2 | 181 | 3154777 |