Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133945.1 | NP_598706.1 | 8, 9 | 784 | ||
GenBank mRNA | AK150435.1 | - | 8, 9 | 875 | ||
AK171673.1 | - | 8, 9 | 789 | |||
AK183463.1 | - | 1, 2 | 52 | |||
AK183527.1 | - | 3, 4 | 167 | |||
BC010473.1 | - | 7, 8 | 767 | 3586304 | ||
BC024782.1 | - | 7, 8 | 767 | 5123949 | ||
BC024839.1 | - | 8, 9 | 784 | 5355521 | ||
BC027129.1 | - | 7, 8 | 713 | |||
BC034196.1 | - | 7, 8 | 765 | 4985176 |