Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300914.1 | NP_001287843.1 | 3 | 1140 | ||
NM_001313964.1 | NP_001300893.1 | 4 | 1066 | |||
NM_006145.2 | NP_006136.1 | 3 | 1289 | |||
XM_006722733.1 | XP_006722796.1 | 4 | 1079 | |||
XM_006722734.3 | XP_006722797.1 | 4 | 1503 | |||
XM_011527956.2 | XP_011526258.1 | 4 | 1058 | |||
GenBank mRNA | AK092989.1 | - | 2 | 355 | ||
AK296609.1 | - | 3 | 674 | |||
AK301817.1 | - | 4 | 981 | |||
BC002352.2 | - | 3 | 1113 | 2821008 | ||
BC019827.1 | - | 3 | 1103 | 4152608 | ||
D49547.1 | - | 3 | 1119 | |||
X62421.1 | - | 3 | 1124 |