Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133967.3 | NP_598728.1 | 8 | 2789 | ||
GenBank mRNA | AK041905.1 | - | 8 | 2658 | ||
AK157968.1 | - | 8 | 2642 | |||
AK170639.1 | - | 8 | 2642 | |||
AK170923.1 | - | 8 | 2614 | |||
AK172462.1 | - | 8 | 2623 | |||
AK196691.1 | - | 1 | 16 | |||
BC013467.1 | - | 7 | 2340 | 4218025 | ||
BC029666.1 | - | 3 | 1743 | |||
BC075666.1 | - | 8 | 2789 | 30605163 |