Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167967.2 | NP_001161439.1 | 4 | 629 | ||
NM_001281974.1 | NP_001268903.1 | 5 | 741 | |||
NM_001281975.1 | NP_001268904.1 | 5 | 791 | |||
NM_133971.3 | NP_598732.2 | 4 | 629 | |||
GenBank mRNA | AK014768.1 | - | 5 | 620 | ||
AK031808.1 | - | 4 | 627 | |||
AK033623.1 | - | 4 | 627 | |||
AK038156.1 | - | 4 | 627 | |||
AK039879.1 | - | 2 | 74 | |||
AK042500.1 | - | 4 | 627 | |||
AK050983.1 | - | 4 | 630 | |||
AK081754.1 | - | 5 | 789 | |||
AK082875.1 | - | 4 | 626 | |||
AK084456.1 | - | 5 | 739 | |||
AK086012.1 | - | 2 | 984 | |||
AK088371.1 | - | 4 | 579 | |||
BC002198.1 | - | 4 | 561 | 3489133 |