Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133993.3 | NP_598754.2 | 5 | 555 | ||
GenBank mRNA | AK009972.1 | - | 5 | 522 | ||
AK011891.1 | - | 5 | 503 | |||
AK075805.1 | - | 5 | 497 | |||
AK082158.1 | - | 5 | 497 | |||
AK149699.1 | - | 5 | 499 | |||
BC003199.1 | - | 5 | 524 | 3586496 | ||
BC023137.1 | - | 5 | 505 | 3986908 |