Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133993.3 | NP_598754.2 | 15 | 1559 | ||
GenBank mRNA | AK003983.1 | - | 5 | 463 | ||
AK009972.1 | - | 15 | 1526 | |||
AK012649.1 | - | 9 | 802 | |||
AK075805.1 | - | 15 | 1500 | |||
AK082158.1 | - | 15 | 1501 | |||
BC003199.1 | - | 15 | 1528 | 3586496 | ||
BC023137.1 | - | 15 | 1509 | 3986908 |