Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133995.4 | NP_598756.1 | 5, 6 | 771 | ||
GenBank mRNA | AK149373.1 | - | 5, 6 | 668 | ||
AK149480.1 | - | 5, 6 | 668 | |||
BC021388.1 | - | 5, 6 | 656 | 5055165 | ||
BC024447.1 | - | 3, 4 | 322 |