Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134012.3 | NP_598773.2 | 9 | 1151 | ||
GenBank mRNA | AK028503.1 | - | 2 | 93 | ||
AK033238.1 | - | 4 | 4024 | |||
AK141155.1 | - | 9 | 1146 | |||
AK153919.1 | - | 11 | 986 | |||
BC020018.1 | - | 5 | 462 | 2646754 | ||
BC062907.1 | - | 9 | 1151 | 6813849 | ||
BC064014.1 | - | 11 | 1255 | 6335954 |