Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134034.2 | NP_598795.2 | 1 | 485 | ||
GenBank mRNA | AK037253.1 | - | 1 | 417 | ||
AK041513.1 | - | 1 | 430 | |||
AK044742.1 | - | 1 | 426 | |||
AK090014.1 | - | 1 | 461 | |||
AK129344.1 | - | 1 | 396 | |||
AK165940.1 | - | 1 | 409 | |||
AK166627.1 | - | 1 | 411 | |||
BC006870.1 | - | 1 | 396 | 3595439 | ||
CD350450.1 | - | 1 | 485 |