Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159536.1 | NP_001153008.1 | 2 | 608 | ||
NM_001159537.1 | NP_001153009.1 | 2 | 667 | |||
NM_138305.3 | NP_612178.2 | 1 | 285 | |||
GenBank mRNA | AF458089.1 | - | 1 | 257 | ||
AK122298.1 | - | 2 | 579 | |||
BC057113.1 | - | 2 | 596 | 6835448 | ||
BC057316.1 | - | 2 | 497 | 6830961 |