Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gldc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138595.2 | NP_613061.1 | 8 | 1278 | ||
GenBank mRNA | AK050981.1 | - | 2 | 109 | ||
AK080789.1 | - | 8 | 1250 | |||
AK080937.1 | - | 8 | 1267 | |||
AK139929.1 | - | 8 | 1269 | |||
BC017135.1 | - | 8 | 1255 | 4511573 |