Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126181.1 | NP_001119653.1 | 4 | 1154 | ||
NM_006176.2 | NP_006167.1 | 4 | 1151 | |||
GenBank mRNA | BC002835.2 | - | 4 | 1042 | 3660038 | |
U89165.1 | - | 4 | 1139 | |||
Y09689.1 | - | 4 | 1128 |