Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251833.1 | NP_001238762.1 | 2 | 624 | ||
NM_001251834.1 | NP_001238763.1 | 2 | 624 | |||
NM_139141.4 | NP_631880.2 | 2 | 624 | |||
GenBank mRNA | AK011044.1 | - | 1 | 444 | ||
AK011096.1 | - | 1 | 444 | |||
AK032718.1 | - | 2 | 580 | |||
AK134299.1 | - | 2 | 609 | |||
BB577095.2 | - | 609 | ||||
BC050152.1 | - | 2 | 560 | |||
BC145448.1 | - | 1 | 458 | 9054006 | ||
BC150775.1 | - | 1 | 458 | 9087686 | ||
CX566615.1 | - | 2 | 583 |