Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2210407C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144544.2 | NP_653127.2 | 4 | 342 | ||
GenBank mRNA | AA124355.1 | - | 4 | 328 | ||
AA434919.1 | - | 2 | 148 | |||
AY148487.1 | - | 3 | 248 | |||
BC019553.1 | - | 4 | 320 | 4456658 | ||
BY096725.1 | - | 4 | 335 | |||
BY097425.1 | - | 4 | 343 |