Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144796.4 | NP_659045.2 | 2 | 355 | ||
GenBank mRNA | AK035779.1 | - | 2 | 355 | ||
AK088629.1 | - | 2 | 268 | |||
BC021842.1 | - | 2 | 365 | 5135798 |