Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006183.4 | NP_006174.1 | 4 | 486 | ||
GenBank mRNA | AF157682.1 | - | 4 | 462 | ||
AK312066.1 | - | 4 | 486 | |||
BC010918.1 | - | 4 | 488 | 4285844 | ||
CR541936.1 | - | 4 | 377 | |||
U91618.1 | - | 4 | 405 |