Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145510.1 | NP_663485.1 | 2 | 430 | ||
GenBank mRNA | AK087547.1 | - | 2 | 425 | ||
AK133528.1 | - | 2 | 451 | |||
AK155054.1 | - | 2 | 430 | |||
AK156126.1 | - | 342 | ||||
AK158129.1 | - | 443 | ||||
AK165748.1 | - | 2 | 448 | |||
BC011166.1 | - | 2 | 430 | 4206804 | ||
BC024808.1 | - | 2 | 420 | 5356768 |