Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ice2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145618.3 | NP_663593.2 | 7 | 919 | ||
GenBank mRNA | AK134186.1 | - | 7 | 964 | ||
AK163917.1 | - | 7 | 847 | |||
AY244558.1 | - | 7 | 948 | |||
BC068183.1 | - | 7 | 919 | 30639735 |