Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145921.1 | NP_666033.1 | 7 | 910 | ||
GenBank mRNA | AK087571.1 | - | 7 | 930 | ||
AK136031.1 | - | 8 | 1166 | |||
BC025001.1 | - | 7 | 910 | 5252879 |