Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145922.2 | NP_666034.1 | 4 | 2042 | ||
GenBank mRNA | AK158073.1 | - | 4 | 1442 | ||
BC024837.1 | - | 4 | 2042 | 5358950 |