Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043344.1 | - | 5, 6 | 693 | ||
AK145200.1 | - | 6, 7 | 749 | |||
AK161473.1 | - | 6, 7 | 832 | |||
AK166305.1 | - | 2, 3 | 258 | |||
BC030328.1 | - | 6, 7 | 749 | 5376020 |