Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290725.1 | NP_001277654.1 | 7 | 1170 | ||
NM_146154.3 | NP_666266.1 | 7 | 1173 | |||
GenBank mRNA | AK032022.1 | - | 5 | 983 | ||
AX775983.1 | - | 7 | 1138 | |||
BC025479.1 | - | 7 | 1135 | 5354224 |