Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146175.3 | NP_666287.3 | 3 | 721 | ||
GenBank mRNA | AK030516.1 | - | 3 | 629 | ||
AK081854.1 | - | 3 | 620 | |||
AK088531.1 | - | 3 | 720 | |||
AK148038.1 | - | 3 | 707 | |||
BC025581.1 | - | 3 | 623 | 5325029 |