Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146207.2 | NP_666319.2 | 8, 9 | 885 | ||
GenBank mRNA | AK144802.1 | - | 8, 9 | 867 | ||
AK150237.1 | - | 8, 9 | 864 | |||
AK168215.1 | - | 8, 9 | 867 | |||
AK170722.1 | - | 8, 9 | 867 | |||
AK196574.1 | - | 1, 2 | 55 | |||
AK201678.1 | - | 1, 2 | 55 | |||
BC010211.1 | - | 8, 9 | 863 | 3969515 | ||
BC024113.1 | - | 4, 5 | 357 | 5097944 | ||
BC027006.1 | - | 4, 5 | 405 |