Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146207.2 | NP_666319.2 | 20 | 3285 | ||
GenBank mRNA | AK003679.1 | - | 4 | 1485 | ||
AK005426.1 | - | 5 | 1588 | |||
AK144802.1 | - | 20 | 3269 | |||
AK168215.1 | - | 20 | 3267 | |||
AK170722.1 | - | 20 | 3267 | |||
BC010211.1 | - | 20 | 3263 | 3969515 | ||
BC024667.1 | - | 2 | 1271 |