Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148930.3 | NP_683732.1 | 14 | 1337 | ||
GenBank mRNA | AJ309168.1 | - | 13 | 1172 | ||
AK134183.1 | - | 14 | 1332 | |||
AK146880.1 | - | 14 | 1332 | |||
BC005719.1 | - | 9 | 942 | |||
BC019766.1 | - | 14 | 1282 | |||
BC023854.1 | - | 14 | 1322 | 5346780 | ||
BC031899.1 | - | 14 | 1322 | 5362121 | ||
BC042419.1 | - | 12 | 4573 | |||
BC043058.1 | - | 14 | 1274 | 6419695 | ||
BC054729.1 | - | 14 | 1325 | 6419758 | ||
BC054820.1 | - | 12 | 4051 | |||
BC060985.1 | - | 13 | 1241 |