Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148930.3 | NP_683732.1 | 17 | 1551 | ||
GenBank mRNA | AJ309168.1 | - | 16 | 1386 | ||
AK040790.1 | - | 1 | 133 | |||
AK083714.1 | - | 1 | 39 | |||
AK134183.1 | - | 17 | 1546 | |||
AK146880.1 | - | 17 | 1546 | |||
BC003988.1 | - | 7 | 500 | |||
BC005719.1 | - | 12 | 1156 | |||
BC019766.1 | - | 17 | 1496 | |||
BC023854.1 | - | 17 | 1536 | 5346780 | ||
BC031899.1 | - | 17 | 1536 | 5362121 | ||
BC042419.1 | - | 15 | 4787 | |||
BC043058.1 | - | 17 | 1488 | 6419695 | ||
BC054729.1 | - | 17 | 1539 | 6419758 | ||
BC054820.1 | - | 15 | 4265 | |||
BC060985.1 | - | 16 | 1455 |