Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152802.3 | NP_690015.1 | 3 | 427 | ||
GenBank mRNA | AB089182.1 | - | 3 | 427 | ||
AK020311.1 | - | 3 | 425 |