Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153119.3 | NP_694759.1 | 4 | 486 | ||
GenBank mRNA | AK028069.1 | - | 4 | 477 | ||
AK154092.1 | - | 4 | 381 | |||
AK158288.1 | - | 4 | 454 | |||
AK170479.1 | - | 4 | 470 | |||
BC028908.1 | - | 4 | 445 | 3668775 |