Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153126.2 | NP_694766.1 | 4 | 507 | ||
GenBank mRNA | AK036294.1 | - | 4 | 455 | ||
AK040070.1 | - | 4 | 462 | |||
AK048552.1 | - | 3 | 332 | |||
AK054029.1 | - | 4 | 528 | |||
AK087445.1 | - | 4 | 465 | |||
AK142682.1 | - | 4 | 519 | |||
AK149825.1 | - | 4 | 334 | |||
BC034516.1 | - | 4 | 507 | 4458024 |