Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153126.2 | NP_694766.1 | 4 | 520 | ||
GenBank mRNA | AK036294.1 | - | 4 | 468 | ||
AK040070.1 | - | 4 | 475 | |||
AK048552.1 | - | 3 | 345 | |||
AK054029.1 | - | 4 | 541 | |||
AK087445.1 | - | 4 | 478 | |||
AK142682.1 | - | 4 | 532 | |||
AK149825.1 | - | 4 | 347 | |||
BC034516.1 | - | 4 | 520 | 4458024 |