Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153168.2 | NP_694808.1 | 21 | 3162 | ||
GenBank mRNA | AJ428066.1 | - | 21 | 3162 | ||
AK032695.1 | - | 21 | 3167 | |||
AK037012.1 | - | 21 | 3171 | |||
AK053667.1 | - | 21 | 3157 | |||
BC053916.1 | - | 22 | 3038 | |||
BC080303.1 | - | 21 | 3049 | 5695617 |