Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153516.2 | NP_705736.1 | 5, 6 | 619 | ||
GenBank mRNA | AK031018.1 | - | 5, 6 | 568 | ||
AK047223.1 | - | 5, 6 | 621 | |||
AK088417.1 | - | 5, 6 | 634 | |||
AK132774.1 | - | 5, 6 | 628 | |||
AK143457.1 | - | 5, 6 | 632 | |||
AK148007.1 | - | 5, 6 | 638 | |||
AK165068.1 | - | 5, 6 | 612 | |||
AK168833.1 | - | 5, 6 | 609 | |||
AK178590.1 | - | 4, 5 | 392 | |||
AK186867.1 | - | 4, 5 | 393 | |||
AK187777.1 | - | 392 | ||||
BC027307.1 | - | 5, 6 | 509 | 4014844 | ||
BC029016.1 | - | 5, 6 | 619 | 5358785 |