Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam102a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153560.4 | NP_705788.1 | 6 | 1296 | ||
GenBank mRNA | AK049032.1 | - | 6 | 1300 | ||
AK131145.1 | - | 5 | 3014 | |||
AK157331.1 | - | 6 | 926 | |||
AK157490.1 | - | 7 | 718 | |||
AK158435.1 | - | 6 | 924 | |||
AK165751.1 | - | 6 | 926 | |||
AK170536.1 | - | 6 | 1324 | |||
AK170847.1 | - | 6 | 917 | |||
AK170951.1 | - | 6 | 923 | |||
AK172536.1 | - | 6 | 923 | |||
BC023752.1 | - | 6 | 741 | 5354455 | ||
BC031157.1 | - | 6 | 559 | 4976773 |