Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam102a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153560.4 | NP_705788.1 | 11 | 3732 | ||
GenBank mRNA | AK049032.1 | - | 11 | 3736 | ||
AK131145.1 | - | 10 | 5450 | |||
AK157331.1 | - | 11 | 3364 | |||
AK157490.1 | - | 12 | 3153 | |||
AK165751.1 | - | 11 | 3386 | |||
AK170536.1 | - | 11 | 3760 | |||
AK192553.1 | - | 1 | 315 | |||
AK198304.1 | - | 1 | 95 | |||
AK203351.1 | - | 1 | 315 | |||
BC023470.1 | - | 3 | 2065 | |||
BC023752.1 | - | 11 | 3177 | 5354455 | ||
BC024513.1 | - | 1 | 463 | |||
BC031157.1 | - | 11 | 2995 | 4976773 |