Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153565.2 | NP_705793.1 | 4 | 969 | ||
GenBank mRNA | AK043077.1 | - | 4 | 819 | ||
AK149520.1 | - | 4 | 748 | |||
AY273821.1 | - | 4 | 593 | |||
BC038085.1 | - | 4 | 984 | 4502199 |